missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRRC4B (aa 508-570) Control Fragment Recombinant Protein

Catalog No. RP101924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101924 Supplier Invitrogen™ Supplier No. RP101924
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63638 (PA5-63638. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q9NT99
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 94030
Name Human LRRC4B (aa 508-570) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A830007M12; HSM; leucine rich repeat containing 4 B; leucine-rich repeat-containing protein 4 B; leucine-rich repeats and immunoglobulin-like domains 4; Lrig4; Lrrc4b; Netrin-G3 ligand; NGL-3; protein NGL-3
Common Name LRRC4B
Gene Symbol LRRC4B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GTEKEPPGPTTDGVWGGGRPGDAAGPASSSTTAPAPRSSRPTEKAFTVPITDVTENALKDLDD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less