missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRP4 (aa 1757-1877) Control Fragment Recombinant Protein

Catalog No. RP89664
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89664 Supplier Invitrogen™ Supplier No. RP89664

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82457 (PA5-82457. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.

Specifications

Accession Number O75096
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4038
Name Human LRP4 (aa 1757-1877) Control Fragment
Quantity 100 μL
Gene Alias 6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2
Common Name LRP4
Gene Symbol LRP4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PGMGNLTYSNPSYRTSTQEVKIEAIPKPAMYNQLCYKKEGGPDHNYTKEKIKIVEGICLLSGDDAEWDDLKQLRSSRGGLLRDHVCMKTDTVSIQASSGSLDDTETEQLLQEEQSECSSVH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less