missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LHFPL2 (aa 155-179) Control Fragment Recombinant Protein

Catalog No. RP91158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91158 Supplier Invitrogen™ Supplier No. RP91158

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59967 (PA5-59967. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LHPL2 is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes.

Specifications

Accession Number Q6ZUX7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10184
Name Human LHFPL2 (aa 155-179) Control Fragment
Quantity 100 μL
Gene Alias 6030465B15; AI447312; AW050335; I79_004617; KIAA0206; LHFPL tetraspan subfamily member 2 protein; Lhfpl2; LHFP-like protein 2; lipoma HMGIC fusion partner-like 2; lipoma HMGIC fusion partner-like 2 protein; mKIAA0206; vgim
Common Name LHFPL2
Gene Symbol LHFPL2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PAGWGCQKAIDYCGHYASAYKPGDC
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less