missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LEPROT (aa 48-68) Control Fragment Recombinant Protein

Catalog No. RP107692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107692 Supplier Invitrogen™ Supplier No. RP107692

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67050 (PA5-67050. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Negatively regulates leptin receptor (LEPR) cell surface expression, and thus decreases response to leptin. Negatively regulates growth hormone (GH) receptor cell surface expression in liver. May play a role in liver resistance to GH during periods of reduced nutrient availability.

Specifications

Accession Number O15243
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54741
Name Human LEPROT (aa 48-68) Control Fragment
Quantity 100 μL
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 6; endospanin-1; LEPR; LEPROT; leptin receptor gene related protein; leptin receptor gene-related protein; leptin receptor overlapping transcript; leptin receptor overlapping transcript protein; OBR; OB-R gene-related protein; OBRGRP; OB-RGRP; VPS55
Common Name LEPROT
Gene Symbol LEPROT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PHFIAKRVTYDSDATSSACRE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less