missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KRT75 (aa 62-108) Control Fragment Recombinant Protein

Catalog No. RP108076
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108076 Supplier Invitrogen™ Supplier No. RP108076

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67414 (PA5-67414. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells.

Specifications

Accession Number O95678
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9119
Name Human KRT75 (aa 62-108) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4732468K03Rik; AA589387; CK-75; cytokeratin KRT2-6 HF; cytokeratin type II; cytokeratin-75; hK6hf; K6HF; K75; KB18; keratin 75; keratin 75, type II; keratin, type II cytoskeletal 75; Keratin-6 hair follicle; keratin-75; keratinocyte associated protein 1; Krt2-6 hf; KRT75; Krtcap1; mK6hf; PFB; type II keratin Kb18; type II keratin-18; type II keratin-K6hf; Type-II keratin Kb18
Common Name KRT75
Gene Symbol KRT75
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LYNLGGAKRVSINGCGSSCRSGFGGRASNRFGVNSGFGYGGGVGGGF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less