missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KLHL7 (aa 188-319) Control Fragment Recombinant Protein

Catalog No. RP92693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92693 Supplier Invitrogen™ Supplier No. RP92693
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56168 (PA5-56168. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Defects in KLHL7 are the cause of retinitis pigmentosa type 42 (RP42). The specific function of this protein remains unknown.

Specifications

Accession Number Q8IXQ5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55975
Name Human KLHL7 (aa 188-319) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2700038B03Rik; BTB/POZ domain containing protein; D5Ertd363e; kelch like family member 7; kelch/BTB; kelch-like 6; kelch-like 7; kelch-like family member 7; kelch-like protein 7; KLHL6; Klhl7; LOW QUALITY PROTEIN: kelch-like protein 7; SBBI26
Common Name KLHL7
Gene Symbol KLHL7
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KRVTHLLNQDTLTVRAEDQVYDAAVRWLKYDEPNRQPFMVDILAKVRFPLISKNFLSKTVQAEPLIQDNPECLKMVISGMRYHLLSPEDREELVDGTRPRRKKHDYRIALFGGSQPQSCRYFNPKDYSWTDI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less