missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KLF2 Alexa Fluor 350-conjugated Antibody, R&D Systems™
Description
KLF2 Monoclonal specifically detects KLF2 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | KLF2 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 665333 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 350 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | Q9Y5W3 |
| Gene Alias | Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor |
| Show More |
For Research Use Only (RUO)