missing translation for 'onlineSavingsMsg'
Learn More

Human KLF12 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. IC9097G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC9097G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC9097G100 Supplier Bio-Techne Supplier No. IC9097G100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Monoclonal Antibody

KLF12 Monoclonal specifically detects KLF12 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen KLF12
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 1230C
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9Y4X4
Gene Alias AP-2rep, AP2REP, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived human KLF12 Asn2-Val402 Accession # Q9Y4X4
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11278
Test Specificity Detects human KLF12 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)