missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KIF12 (aa 555-646) Control Fragment Recombinant Protein

Catalog No. RP92968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92968 Supplier Invitrogen™ Supplier No. RP92968

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (63%), Rat (63%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54161 (PA5-54161. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

KIF12 encodes a member of the kinesin superfamily of microtubule-associated molecular motors with functions related to the microtubule cytosekelton. Members of this superfamily play important roles in intracellular transport and cell division. A similar protein in mouse functions in the beta cell antioxidant signaling cascade, acting as a scaffold for the transcription factor specificity protein 1 (Sp1). Mice that lack this gene exhibit beta cell oxidative stress resulting in hypoinsulinemic glucose intolerance.

Specifications

Accession Number Q96FN5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 113220
Name Human KIF12 (aa 555-646) Control Fragment
Quantity 100 μL
Gene Alias KIF12; kinesin 12; kinesin family member 12; kinesin superfamily protein 12; kinesin-like protein KIF12; N-9 kinesin; RP1156P10\0.3
Common Name KIF12
Gene Symbol KIF12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AKCPRERSHSDWTQTRVLAEMLTEEEVVPSAPPLPVRPPKTSPGLRGGAGVPNLAQRLEALRDQIGSSLRRGRSQPPCSEGARSPGQVLPPH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less