missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KARS (aa 481-591) Control Fragment Recombinant Protein

Catalog No. RP98537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98537 Supplier Invitrogen™ Supplier No. RP98537
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59620 (PA5-59620. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Lysyl-tRNA synthetase is a homodimer localized to the cytoplasm which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Accession Number Q15046
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3735
Name Human KARS (aa 481-591) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; EC 6.1.1.6; KARS; Kars1; KARS2; KIAA0070; KRS; lysine tRNA ligase; lysine--tRNA ligase; LysRS; lysyl-tRNA synthetase; mKIAA0070
Common Name KARS
Gene Symbol KARS1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GLTERFELFVMKKEICNAYTELNDPMRQRQLFEEQAKAKAAGDDEAMFIDENFCTALEYGLPPTAGWGMGIDRVAMFLTDSNNIKEVLLFPAMKPEDKKENVATTDTLEST
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less