missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Jak3 Alexa Fluor 700-conjugated Antibody, R&D Systems™
Description
Jak3 Monoclonal specifically detects Jak3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | Jak3 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 452524 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | P52333 |
| Gene Alias | EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKL, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte) |
| Host Species | Mouse |
| Show More |
For Research Use Only (RUO)