missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Islet-1 ELISA Kit

Catalog No. EH280RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH280RBX5 5 x 96 Tests
EH280RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH280RBX5 Supplier Invitrogen™ Supplier No. EH280RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ELISA

Human Islet-1 quantitates human Islet-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Islet-1.

This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.

Specifications

Accession Number P61371
Assay Range 0.1-25 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Insulin gene enhancer protein ISL-1; insulin related protein; ISL LIM homeobox 1; Isl1; Isl-1; isl-1 homeobox; ISL1 transcription factor LIM/homeodomain (islet-1); ISL1 transcription factor, LIM/homeodomain; ISL1 transcription factor, LIM/homeodomain 1; isl-1=homeobox; Islet 1; ISLET1; islet-1; Lim-homeodomain protein Islet1Insulin gene enhancer protein ISL-1; insulin related protein; ISL LIM homeobox 1; Isl1; Isl-1; isl-1 homeobox; ISL1 transcription factor LIM/homeodomain (islet-1); ISL1 transcription factor, LIM/homeodomain; ISL1 transcription factor, LIM/homeodomain 1; isl-1=homeobox; Islet 1; ISLET1; islet-1; Lim-homeodomain protein Islet1;
Gene ID (Entrez) 3670
Gene Symbol ISL1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target ISL1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less