missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human INTS4 (aa 8-110) Control Fragment Recombinant Protein

Catalog No. RP91646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91646 Supplier Invitrogen™ Supplier No. RP91646
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59962 (PA5-59962. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.

Specifications

Accession Number Q96HW7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 92105
Name Human INTS4 (aa 8-110) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610034N24Rik; AA692684; AW493223; int4; Integrator complex subunit 4; Ints4; MST093; MSTP093; RGD1305036
Common Name INTS4
Gene Symbol INTS4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RVYEEFTKVVQPQEEIATKKLRLTKPSKSAALHIDLCKATSPADALQYLLQFARKPVEAESVEGVVRILLEHYYKENDPSVRLKIASLLGLLSKTAGFSPDCI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less