missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL1RAPL2 (aa 226-303) Control Fragment Recombinant Protein

Catalog No. RP95474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP95474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP95474 Supplier Invitrogen™ Supplier No. RP95474

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57530 (PA5-57530. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the interleukin 1 receptor family. This protein is similar to the interleukin 1 accessory proteins, and is most closely related to interleukin 1 receptor accessory protein-like 1 (IL1RAPL1). This gene and IL1RAPL1 are located at a region on chromosome X that is associated with X-linked non-syndromic mental retardation. [provided by RefSeq, Jul 2008].

Specifications

Accession Number Q9NP60
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26280
Name Human IL1RAPL2 (aa 226-303) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias IL-1 receptor accessory protein-like 2; IL1R9; IL-1R9; IL-1 R-9; IL1RAPL2; IL-1-RAPL-2; IL-1 RAPL-2; IL1RAPL-2; IL1RAPL-2-related protein; interleukin 1 receptor 9; interleukin 1 receptor accessory protein like 2; interleukin 1 receptor accessory protein-like 2; Interleukin-1 receptor 9; RGD1561761; three immunoglobulin domain-containing IL-1 receptor-related 1; TIGIRR-1; X-linked interleukin-1 receptor accessory protein-like 2
Common Name IL1RAPL2
Gene Symbol IL1RAPL2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TTELKVTALLTDKPPKPLFPMENQPSVIDVQLGKPLNIPCKAFFGFSGESGPMIYWMKGEKFIEELAGHIREGEIRLL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less