missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-7 Recombinant Protein, PeproTech®

Catalog No. p-7246297
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20007100UG 100 μg
2000710UG 10 μg
2000701M 2 x 500 μg
200071MG 2 x 500 μg
20007250UG 250 μg
200072UG 2 μg
20007500UG 500 μg
2000750UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 20007100UG Supplier Gibco™ Supplier No. LS20007100UG
Only null left

Recombinant Protein

200-07-1MG will be provided as 2 x 500 μg (200-07-500UG). Recombinant Human IL-7 is a 17.5 kDa protein containing 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.

Specifications

Accession Number P13232, P16871
For Use With (Application) Control, Western Blot Control, ELISA Standard, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3574, 3575
Molecular Weight (g/mol) 17.5 kDa
Name Human IL-7
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7
Common Name IL-7
Gene Symbol Il7, IL7r
Biological Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of murine 2E8 cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH
Content And Storage -20°C
Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of murine 2E8 cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less