missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-7 Recombinant Protein DFS Item

Recombinant Protein

$388.00 - $10250.00

Specifications

Accession Number P13232, P16871
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3574, 3575
Molecular Weight (g/mol) 17.5 kDa
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC0071 DFS Item
View Documents
Gibco™
PHC0071
100 μg
Each for $2,025.00
Only null left
 
PHC0073 DFS Item
View Documents
Gibco™
PHC0073
1 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PHC0075 DFS Item
View Documents
Gibco™
PHC0075
5 μg
Each for $388.00
Only null left
 
PHC0076 DFS Item
View Documents
Gibco™
PHC0076
25 μg
Each for $1,184.00
Only null left
 
Description

Description

Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water to 0.1-1.0 mg/mL to regain full activity. Apportion the reconstituted protein into working aliquots and store at ≤-20°C. Make any further dilutions of the reconstituted protein in low endotoxin medium or buffered solution with FBS or tissue culture grade BSA. Storage: Store the lyophilized protein at 2-8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C (not in a frost-free freezer). Avoid repeated freeze-thaw cycles.

Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
Specifications

Specifications

P13232, P16871
Protein with no preservative
17.5 kDa
Purified
RUO
A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7
Il7, IL7r
Protein
Human
DCDIEGKDGK QYESVLMVSI DQLLDSMKEI GSNCLNNEFN FFKRHICDAN KEGMFLFRAA RKLRQFLKMN STGDFDLHLL KVSEGTTILL NCTGQVKGRK PAALGEAQPT KSLEENKSLK EQKKLNDLCF LKRLLQEIKT CWNKILMGTK EH
Carrier-Free
<0.1 ng/μg
E. coli
Recombinant, Ligand
Interleukins
Gibco
Bioactivity
3574, 3575
Human IL-7
4°C
<0.1 ng/μg
IL-7
ED50 = 0.2 - 1 ng/mL; determined by the dose-dependent proliferation of human PBMCs.
Unconjugated
Recombinant
4°C
ED50 = 0.2 - 1 ng/mL; determined by the dose-dependent proliferation of human PBMCs.
Approved for shipment on Wet or Dry Ice
Cytokines & Receptors
Lyophilized
Immunology, Signal Transduction, Stem Cell Research, Drug Development, Oncology
>95% by SDS-PAGE
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'