missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-6 Recombinant Protein, PeproTech®
Recombinant Protein
$131.00 - $4,870.00
Specifications
| Accession Number | P05231 |
|---|---|
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3569 |
| Molecular Weight (g/mol) | 20.9 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
20006100UG
|
Gibco™
LS20006100UG |
100 μg |
N/A
|
|
|||||
200061MG
|
Gibco™
LS200061MG |
1 mg |
N/A
|
|
|||||
2000620UG
|
Gibco™
LS2000620UG |
20 μg |
N/A
|
|
|||||
20006250UG
|
Gibco™
LS20006250UG |
250 μg |
N/A
|
|
|||||
20006500UG
|
Gibco™
LS20006500UG |
500 μg |
N/A
|
|
|||||
2000650UG
|
Gibco™
LS2000650UG |
50 μg |
N/A
|
|
|||||
200065UG
|
Gibco™
LS200065UG |
5 μg |
N/A
|
|
|||||
Description
Recombinant Human IL-6 is a 20.9 kDa protein containing 184 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.Specifications
| P05231 | |
| protein with no preservative | |
| 20.9 kDa | |
| E. coli | |
| <1 EU/ μg | |
| IL-6 | |
| Determined by its ability to stimulate the proliferation of mouse B9 cells. | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| ELISA, Functional Assay, Western Blot | |
| 3569 | |
| Human IL-6 | |
| RUO | |
| B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 | |
| Il6 | |
| Unconjugated | |
| PVPPGEDSKD VAAPHRQPLT SSERIDKQIR YILDGISALR KETCNKSNMC ESSKEALAEN NLNLPKMAEK DGCFQSGFNE ETCLVKIITG LLEFEVYLEY LQNRFESSEE QARAVQMSTK VLIQFLQKKA KNLDAITTPD PTTNASLLTK LQAQNQWLQD MTTHLILRSF KEFLQSSLRA LRQM | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |