Learn More
Invitrogen™ Human IL-6 Uncoated ELISA Kit with Plates
ELISA
Supplier: Invitrogen™ 88706622
Description
The Human Interleukin-6 (IL-6) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-6. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.Chemical Identifiers
| P05231 | |
| 2 pg/mL | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| CVID11,IL-21,Za11 | |
| HRP | |
| Plasma 100 μL, Serum 100 μL, Supernatant 100 μL | |
| Interleukin-6 | |
| 1 hrs 45 mins |
| 2 to 200 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| 3569 | |
| Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86) | |
| RUO | |
| 2°C to 8°C | |
| IL-6 | |
| 24 hrs 30 mins |
Specifications
| P05231 | |
| 2 pg/mL | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| CVID11,IL-21,Za11 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| IL-6 | |
| 24 hrs 30 mins |
| 2 to 200 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| 3569 | |
| Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86) | |
| 2 x 96 Tests | |
| Plasma 100 μL, Serum 100 μL, Supernatant 100 μL | |
| Interleukin-6 | |
| 1 hrs 45 mins |