missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-6 ELISA Kit

Catalog No. ENEH2IL6
Encompass
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENEH2IL6 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENEH2IL6 Supplier Invitrogen™ Supplier No. EH2IL6

ELISA

The Human Interleukin-6 (Hu IL-6) ELISA quantitates Hu IL-6 in human serum, plasma, urine, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-6. Principle of the method The Human IL-6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.

Specifications

Accession Number P05231
Assay Range 10.24 to 400 pg/mL
Assay Sensitivity <1 pg/mL
Description Human IL-6 ELISA Kit
Conjugate HRP
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Product Type ELISA
Sample Type Plasma, Serum, Supernatant, Urine
Specificity No significant cross-reaction with with human IL-1 A, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-10, IL-12, TNFa, IFNg, IFNa, GM-CSF, or mouse IL-6
Sufficient For 96 microplate wells
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3569
Gene Symbol BSF-2,BSF2,CDF,HGF,HSF,IFN-beta-2,IFNB2,IL-6
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Measurement Absorbance at 450 nm minus absorbance (at 550 nm)
Precision Intra-assay CV <10%; inter-assay CV <10%
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 50 μL
Storage Requirements 2°C to 8°C
Target IL-6
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs
Show More Show Less