missing translation for 'onlineSavingsMsg'
Learn More

Human IL-4I1 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Catalog No. IC56843S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC56843S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC56843S100 Supplier Bio-Techne Supplier No. IC56843S100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

IL-4I1 Monoclonal specifically detects IL-4I1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen IL-4I1
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 1006231
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q96RQ9
Gene Alias FIG1, Fig-1 protein, hFIG1, IL4I1, IL4-induced protein 1, interleukin 4 induced 1, interleukin four induced 1, LAAO, L-amino-acid oxidase, LAO, UNQ636/PRO1265
Host Species Rat
Immunogen Chinese Hamster Ovary cell line, CHO-derived human IL-4I1, Met1-His567, Accession # Q96RQ9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259307
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line R&D Systems
Show More Show Less