missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-4 ELISA Kit

ELISA

Specifications

Accession Number P05112
Assay Range 7.8-500 pg/mL
Assay Sensitivity 1.3 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-4
Biotin-Conjugate anti-human IL-4 monoclonal antibody
Streptavidin-HRP
Human IL-4 Standard lyophilized, 1ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
5018073
View Documents Promotion Details
Invitrogen™
BMS2252
96 Tests N/A
N/A
Description

Description

The Human Interleukin-4 (Hu IL-4) ELISA quantitates Hu IL-4 in human serum, plasma, urine or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-4. Principle of the method The Human IL-4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
Specifications

Specifications

P05112
1.3 pg/mL
ELISA
Human
Absorbance Microplate Reader
Il4
4.8%
Biotin
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
B-cell stimulatory factor 1, B cell growth factor 1
1 hr 20 min
7.8-500 pg/mL
HRP
Plasma, Serum, Supernatant
ELISA
3565
5.6%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2-8°C
IL-4
3 hr 10 min
Videos
SDS
Documents

Documents