missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-34 (Carrier-Free) Recombinant Protein, eBioscience™

Recombinant Protein

$199.00 - $199.00

Specifications

Accession Number Q6ZMJ4
Formulation 0.02M citric acid with no preservative; pH 3.0
Gene ID (Entrez) 146433
Molecular Weight (g/mol) 52.5kDa
Name Human IL-34 Carrier-Free
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123510
View Documents
Invitrogen™
34868461
2 μg
Each for $199.00
Only null left
 
Description

Description

IL-34 is secreted as a dimeric protein and currently belongs to no known cytokine family. It signals via the M-CSF Receptor, also known as CSF-1R and CD115, which is shared with M-CSF. IL-34 has no amino acid sequence homology to M-CSF, but they exhibit similarities in secondary structure and share overlapping functions. IL-34 has been shown to promote myeloid survival, proliferation, and differentiation, and to play a role in osteoclast formation. IL-34 expression has been detected in various tissues and is most abundant in the spleen. Description: IL-34 is secreted as a dimeric protein and currently belongs to no known cytokine family. It signals via the M-CSF Receptor, also known as CSF-1R and CD115, which is shared with M-CSF. IL-34 has no amino acid sequence homology to M-CSF, but they exhibit similarities in secondary structure and share overlapping functions. IL-34 has been shown to promote myeloid survival, proliferation, and differentiation, and to play a role in osteoclast formation. IL-34 expression has been detected in various tissues and is most abundant in the spleen. Applications Reported: Recombinant human IL-34 is biologically active and can induce MCP-1 secretion by normal human peripheral blood cells. Applications Tested: The activity of this protein was determined by measuring MCP-1 secretion from normal human peripheral blood cells. Source: HEK293 cells (amino acids Asn21-Pro242; accession # NP_001166243); contains a C-terminal 8X His tag. Bioactivity: The activity of this protein was determined by measuring MCP-1 secretion from normal human peripheral blood cells. Endotoxin: Less than 0.1ng/ug cytokine as determined by the LAL assay. Purity: >95% as determined by SDS-PAGE and HPLC. Molecular Weight: 52.5 kDa. Storage and handling: For best recovery, quick spin vial prior to opening. Use in sterile environment. Reconstitute in water to a concentration of 0.1-1.0mg/mL. Do not vortex. Store lyophilized protein at less than or equal to -20°C. Reconstituted protein solution can be stored with carrier protein (e.g., 0.1% BSA) at less than or equal to -20°C.
Specifications

Specifications

Q6ZMJ4
146433
Human IL-34 Carrier-Free
SDS-PAGE and HPLC
RUO
Less than 0.1ng/μg cytokine as determined by the LAL assay
IL34
Recombinant Protein, Carrier-Free
Unconjugated
Human (HEK293)
Lyophilized
Affinity chromatography
0.02M citric acid with no preservative; pH 3.0
52.5kDa
3.0
-20°C
HEK 293 cells (amino acids Asn21-Pro242; Accession # NP_001166243); contains a C-terminal 8X His tag
Human IL-34 Carrier-Free
The activity of this protein was determined by measuring MCP-1 secretion from normal human peripheral blood cells.
Human
Hu
Recombinant
Recombinant
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'