missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-3 Recombinant Protein DFS Item

Recombinant Protein

Supplier:  Gibco™ PHC0033

 View more versions of this product

Catalog No. PHC0033


Only null left
Explore available promotions

Description

Description

Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute lyophilized recombinant human IL-3 in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Stock solutions should be apportioned into working aliquots and stored at ≤-20°C. Further dilution should be made in medium or buffered solution containing carrier protein, such as PBS with 0.1% BSA. Storage: Lyophilized recombinant human IL-3 should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted recombinant human IL-3 at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Specifications

Specifications

P08700
Protein with no preservative
15 kDa
Purified
4°C
<0.1 ng/μg
IL-3
ED50 = 0.1 - 0.25 ng/mL; determined by the dose-dependent proliferation of human TF-1 cells.
Unconjugated
Recombinant
4°C
ED50 = 0.1 - 0.25 ng/mL; determined by the dose-dependent proliferation of human TF-1 cells.
Wet Ice
Cytokines & Receptors
Lyophilized
Stem Cell Research, Immunology
>95% by SDS-PAGE
Bioactivity
3562
Human IL-3
1 mg
RUO
BP/BPA; BPA; Colony stimulating factor multiple; colony-stimulating factor, multiple; Csfmu; Eo-CSF; HCGF; hematopoietic growth factor; Il3; Il-3; ILN; Interleukin; interleukin 3; Interleukin 3 (colony stimulating factor; Interleukin3; interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; MGC79398; MGC79399; MGF; Multi CSF; MULTI-CSF; Multilineage colony stimulating factor; multilineage-colony-stimulating factor; multiple); multipotential colony-stimulating factor; P cell stimulating factor; P-cell stimulating factor; P-cell-stimulating factor; PCSA; PSF
IL3
Protein
Human
APMTQTTPLK TSWVNCSNMI DEIITHLKQP PLPLLDFNNL NGEDQDILME NNLRRPNLEA FNRAVKSLQN ASAIESILKN LLPCLPLATA APTRHPIHIK DGDWNEFRRK LTFYLKTLEN AQAQQTTLSL AIF
Carrier-Free
<0.1 ng/μg
E. coli
Recombinant, Ligand
Interleukins
Gibco
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions