Learn More
Invitrogen™ Human IL-3 ELISA Development Kit (ABTS), PeproTech®
ELISA
Supplier:  Invitrogen™ 900K13K
Catalog No. 90-0K1-3K
Only null left
Explore available promotions
Description
Description
Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Specifications
Specifications
P08700
Lot-specific
ELISA
3562
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
1 hrs 45 mins
Lot-specific
HRP
Colorimetric Microplate Reader
IL3
HRP
RUO
IL-3
24 hrs 30 mins
Product Suggestions
Product Suggestions