missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-21 Proquantum Immunoassay Kit
ELISA
Supplier: Invitrogen™ A35593
Description
IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.Specifications
| Q9HBE4 | |
| <0.3 pg/mL | |
| ProQuantum | |
| qPCR Instruments | |
| IL21 | |
| FAM™ | |
| RUO | |
| -20°C | |
| 30 mins |
| 0.32 to 5000 pg/mL | |
| FAM™ | |
| Plasma, Serum, Supernatant | |
| 59067 | |
| Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
| 96 Tests | |
| Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
| IL-21 | |
| 2 hrs |