missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-2 Recombinant Protein, PeproTech®
Description
Recombinant Human IL-2 is a 15.5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
Specifications
| Accession Number | P60568 |
| For Use With (Application) | Bioactivity, Control, ELISA Standard, Western Blot Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3558 |
| Molecular Weight (g/mol) | 15.5 kDa |
| Name | Human IL-2 |
| Purification Method | Purified |
| Quantity | 50 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |