Learn More
Invitrogen™ Human IL-2 ELISA Development Kit (ABTS), PeproTech®
ELISA
Supplier:  Invitrogen™ 900K12K
Catalog No. 90-0K1-2K
Only null left
Explore available promotions
Description
Description
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
Specifications
P60568
Lot-specific
ELISA
3558
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
1 hrs 45 mins
Lot-specific
HRP
Colorimetric Microplate Reader
IL2
HRP
RUO
IL-2
24 hrs 30 mins
Product Suggestions
Product Suggestions