missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL1RA ELISA Kit

ELISA

$856.00 - $5960.00

Specifications

Accession Number P18510
Assay Range 109 to 7,000 pg/mL
Assay Sensitivity 30 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal polyclonal antibody to human IL-1RA
Biotin-Conjugate anti-human IL-1RA polyclonal antibody
Streptavidin-HRP
Human IL-1RA Standard lyophilized, 14ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5017982
View Documents
Invitrogen™
BMS2080
96 Tests
Each for $856.00
Only null left
 
5017983
View Documents
Invitrogen™
BMS2080TEN
10 x 96 Tests
Each for $5,960.00
Only null left
 
Description

Description

The Human Interleukin-1 Receptor Antagonist (Hu IL-1RA) ELISA quantitates Hu IL-1RA in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-1RA. Principle of the method The Human IL-1RA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
Specifications

Specifications

P18510
30 pg/mL
ELISA
Human
Colorimetric Microplate Reader
DIRA,ICIL-1 RA,IL-1 RN,IL-1 ra,IL-1 ra3,IL1F3,IL1RA,IRAP,MVCD4
7.2
HRP
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Interleukin-1 receptor antagonist
1 hrs 20 mins
109 to 7,000 pg/mL
HRP
Plasma, Serum, Supernatant
ELISA
3557
7.4%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
IL1RA
3 hrs 30 mins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'