missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-1RA, Animal-Free Recombinant Protein, PeproTech®
Description
Recombinant Human IL-1RA is a 17.2 kDa protein consisting of 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
Specifications
Specifications
| Accession Number | P18510 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3557 |
| Molecular Weight (g/mol) | 17.2 kDa |
| Name | Human IL-1 RA, Animal-Free |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 250 μg |
| Endotoxin Concentration | <0.1 EU/ μg |
| Show More |