missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-1 beta ELISA Kit

ELISA

$874.00 - $6,510.00

Specifications

Accession Number P01584
Assay Range 3.9 to 250 pg/mL
Assay Sensitivity 0.3 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-1beta
Biotin-Conjugate anti-human IL-1beta monoclonal antibody
Streptavidin-HRP
Human IL-1beta Standard lyophilized, 500pg/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
5018066
View Documents Promotion Details
Invitrogen™
BMS2242
96 Tests
Each for $874.00
Only null left
 
5018068
View Documents Promotion Details
Invitrogen™
BMS2242TEN
10 x 96 Tests
Each for $6,510.00
Only null left
 
Description

Description

The Human Interleukin-1beta (Hu IL-1β) ELISA quantitates Hu IL-1β in human serum, plasma, urine or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-1β. Principle of the method The Human IL-1β solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
Specifications

Specifications

P01584
0.3 pg/mL
ELISA
Human
Colorimetric Microplate Reader
IL-1,IL1-BETA,IL1F2
5.1
HRP
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 50 μL
Catabolin, Interleukin-1 beta, IL1F2
1 hrs 20 mins
3.9 to 250 pg/mL
HRP
Plasma, Serum, Supernatant, Urine
ELISA
3553
8.6%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Controls, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
IL-1 beta
3 hrs 10 mins
Videos
SDS
Documents

Documents