missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-17F, Animal-Free Recombinant Protein, PeproTech®
Recombinant Protein
$339.00 - $5,010.00
Specifications
| Accession Number | Q96PD4 |
|---|---|
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 112744 |
| Molecular Weight (g/mol) | 30.1 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AF20025100
|
Gibco™
AF20025100UG |
100 μg |
N/A
|
|
|||||
AF200251MG
|
Gibco™
AF200251MG |
1 mg |
N/A
|
|
|||||
AF2002525UG
|
Gibco™
AF2002525UG |
25 μg |
N/A
|
|
|||||
AF20025500
|
Gibco™
AF20025500UG |
500 μg |
N/A
|
|
|||||
Description
The mature human IL-17F is a homodimeric protein with a total weight of 30.1 kDa, consisting of two 133 amino acid residue chains. E.coli-derived Recombinant Human IL-17F is a biologically active, non-glycosylated, disulfide-linked homodimeric protein containing 268 amino acids (30.1 kDa), including N-terminal methionine residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IL-17F (Interleukin 17F, CTLA-8)) is a cytokine belonging to the IL-17 family that is produced by inflammatory cells such as activated T cells, mast cells, and basophils. IL-17F is involved in allergic airway inflammation, and can induce several cytokines, chemokines, and adhesion molecules in bronchial epithelial cells, vein endothelial cells, fibroblasts, and eosinophils. IL-17F may be secreted as a homodimer, or a heterodimer with IL17A. It acts by binding to the type I receptor, IL-17R, aiding recruitment of monocytes and neutrophils at the site of inflammation by increasing chemokine production. IL-17F also stimulates induction of other pro-inflammatory cytokines TNF alpha, IL-1 beta, IL-6, and IL-8, and reports strongly suggest the involvement of IL-17 in several chronic inflammatory diseases such as rheumatoid arthritis, psoriasis and multiple sclerosis. TGF-beta (differentiation) and IL-23 (expansion) are required for induction and maintenance of Th17 (IL-17 producing) cells, which in turn induce the other pro-inflammatory cytokines. IL-17F is produced, and exists, as a homo-dimer, with homology to a herpes virus early protein, is one of the six members (IL-17A-F) of this cytokine family, and is well characterized and highly expressed by activated effector memory T cells. IL-17F has been found to inhibit the angiogenesis of endothelial cells and induce endothelial cells to produce IL2, TGFB1/TGFB, and monocyte chemoattractant protein-1.Specifications
| Q96PD4 | |
| protein with no preservative | |
| 30.1 kDa | |
| E. coli | |
| <0.1 EU/ μg | |
| IL-17 F | |
| Measured by its ability to induce IL-6 production by NHDF cells. | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| Functional Assay | |
| 112744 | |
| Human IL-17 F, Animal-Free | |
| RUO | |
| C87042; CANDF6; cytokine ML-1; IL17F; IL-17 F; IL-24; ILN; Interleukin; interleukin 17 F; Interleukin17F; interleukin-17 F; interleukin-24; ML1; ML-1; mutant IL-17 F | |
| IL17F | |
| Unconjugated | |
| MRKIPKVGHT FFQKPESCPP VPGGSMKLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGKEDISMN SVPIQQETLV VRRKHQGCSV SFQLEKVLVT VGCTCVTPVI HHVQ | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |