missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-17E (IL-25), Animal-Free Recombinant Protein, PeproTech®
Description
Recombinant Human IL-17E is a 33.8 kDa disulfide-linked homodimer of two 146 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
Specifications
Specifications
| Accession Number | Q9H293 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 64806 |
| Molecular Weight (g/mol) | 33.8 kDa |
| Name | Human IL-17 E (IL-25), Animal-Free |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 250 μg |
| Endotoxin Concentration | <0.1 EU/ μg |
| Show More |