missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17E/IL-25 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | IL-17E/IL-25 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 182203 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | Q9H293 |
| Gene Alias | IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25 |
| Show More |
For Research Use Only (RUO)