Learn More
Invitrogen™ Human IL-17AF ELISA Kit
ELISA
$804.00 - $5500.00
Specifications
| Accession Number | Q16552,Q96PD4 |
|---|---|
| Assay Range | 31.3 to 2,000 pg/mL |
| Assay Sensitivity | 8.8 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-17A
Biotin-Conjugate anti-human IL-17F monoclonal antibody
Streptavidin-HRP
Human IL-17AF Standard lyophilized, 2000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Calibrator Diluent
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Human Interleukin-17AF (Hu IL-17AF) ELISA quantitates Hu IL-17AF in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-17AF. Principle of the method The Human IL-17AF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis.Specifications
| Q16552,Q96PD4 | |
| 8.8 pg/mL | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| CANDF6,IL-17 F,ML-1,ML1,Ctla-8,Ctla8,IL-17,IL-17 A,Il17 | |
| 5.9 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Interleukin 17 AF, IL-17 A/F heterodimer | |
| 1 hrs 20 mins |
| 31.3 to 2,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| 112744, 3605 | |
| 5.2% | |
| Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| IL-17 AF | |
| 3 hrs 30 mins |