missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-17AF (heterodimer) Uncoated ELISA Kit with Plates

ELISA

Specifications

Accession Number Q16552,Q96PD4
Assay Range 30 to 4,000 pg/mL
Assay Sensitivity 30 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
5017243
View Documents Promotion Details
Invitrogen™
88711722
2 x 96 Tests N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
5017244
View Documents Promotion Details
Invitrogen™
88711786
10 x 96 Tests N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

The Human IL-17AF (heterodimer) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-17AF. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis.
Specifications

Specifications

Q16552,Q96PD4
30 pg/mL
ELISA
Human
Colorimetric Microplate Reader
IL-5
HRP
Plasma 100 μL, Serum 100 μL, Supernatant 100 μL
Interleukin-IL-17 AF, IL17AF
1 hrs 45 mins
30 to 4,000 pg/mL
HRP
Plasma, Serum, Supernatant
ELISA
112744, 3605
Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
RUO
2°C to 8°C
IL-17 AF
24 hrs 30 mins
Videos
SDS
Documents

Documents