missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17/IL-17A Alexa Fluor 700-conjugated Antibody, R&D Systems™
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | IL-17/IL-17A |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 41802 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 700 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | Q16552 |
| Gene Alias | CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
| Show More |
For Research Use Only (RUO)