Learn More
Invitrogen™ Human IL-13 ELISA Kit
ELISA
$856.00 - $5960.00
Specifications
| Accession Number | P35225 |
|---|---|
| Assay Range | 1.6-100 pg/mL |
| Assay Sensitivity | 0.7 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-13
Biotin-Conjugate anti-human IL-13 monoclonal antibody
Streptavidin-HRP
Human IL-13 Standard lyophilized, 200pg/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
Description
The Human Interleukin-13 (Hu IL-13) ELISA quantitates Hu IL-13 in human serum, plasma, amniotic fluid or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-13. Principle of the method The Human IL-13 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.Specifications
| P35225 | |
| 0.7 pg/mL | |
| ELISA | |
| Human | |
| Absorbance Microplate Reader | |
| Il13 | |
| 6% | |
| Biotin | |
| Amniotic Fluid 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| ALRH, BHR1 | |
| 1 hr 20 min |
| 1.6-100 pg/mL | |
| HRP | |
| Amniotic Fluid, Plasma, Serum, Supernatant | |
| ELISA | |
| 3596 | |
| 4.6% | |
| Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2-8°C | |
| IL-13 | |
| 2 hr 10 min |