Learn More
Invitrogen™ Human IL-12 p70 ELISA Kit
ELISA
$856.00 - $6120.00
Specifications
| Accession Number | P29459,P29460 |
|---|---|
| Assay Range | 3.1 to 200 pg/mL |
| Assay Sensitivity | 2.1 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-12 p70
Biotin-Conjugate anti-human IL-12 p70 monoclonal antibody
Streptavidin-HRP
Human IL-12 p70 Standard lyophilized, 400pg/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Human Interleukin-12 p70 (IL-12 p70) ELISA quantitates Hu IL-12 p70 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-12 p70. Principle of the method The Human IL-12 p70 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.Specifications
| P29459,P29460 | |
| 2.1 pg/mL | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| CLMF,CLMF2,IL-12 B,IMD28,IMD29,NKSF,NKSF2,CLMF,IL-12 A,NFSK,NKSF1,P35 | |
| 3 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Interleukin-12 subunit p70, CLMF, NKSF | |
| 1 hrs 20 mins |
| 3.1 to 200 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| 3592, 3593 | |
| 4.8% | |
| Pre-coated 96 well plate, Adhesive Plate Covers, Sample Diluent, Assay Buffer concentrate, Assay Buffer concentrate, SAV-HRP, SAV-HRP, Controls, Controls, Chromogen, Stop Solution | |
| RUO | |
| 2°C to 8°C | |
| IL-12 p70 | |
| 3 hrs 10 mins |