missing translation for 'onlineSavingsMsg'
Learn More

Human IL-12/IL-35 p35 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Catalog No. IC15701V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC15701V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC15701V100 Supplier Bio-Techne Supplier No. IC15701V100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Monoclonal Antibody

IL-12/IL-35 p35 Monoclonal specifically detects IL-12/IL-35 p35 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen IL-12/IL-35 p35
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 2038C
Conjugate Alexa Fluor 405
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P29460 (p40) & P29459 (P35)
Host Species Rabbit
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 p40 and human IL-12 p35 Ile23-Ser328 of p40, Arg23-Ser219 of p35 Accession # P29460 (p40) & P29459 (P35)
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human IL-12/IL-35 p35 in direct ELISAs. Detects the IL-12/IL-35 p35 subunit either as part of the IL-12 p40/p35 heterodimer or as the IL-12/IL-35 p35 monomer. This antibody does not detect the IL-12/IL-23 p40 subunit.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Form Supernatant
Isotype IgG
Product Line R&D Systems
Show More Show Less