missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-11 Recombinant Protein DFS Item

Recombinant Protein

Supplier:  Gibco™ PHC0115

Catalog No. PHC0115


Only null left
Explore available promotions

Description

Description

Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute recombinant human IL-11 in sterile, distilled water to a concentration of 0.5-1.0 mg/mL. It is important to note that this protein is slow to dissolve. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage, preferably desiccated. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.

Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.
Specifications

Specifications

P20809
Protein with no preservative
19.1 kDa
Purified
-20°C
<0.1 ng/μg
IL-11
ED50 ≤ 2 ng/mL; determined by the dose-dependent proliferation of murine T11 cells.
Unconjugated
Recombinant
-20°C
ED50 ≤ 2 ng/mL; determined by the dose-dependent proliferation of murine T11 cells.
Approved for shipment on Wet or Dry Ice
Cytokines & Receptors
Lyophilized
Signal Transduction, Stem Cell Research, Bone Research
≥98% by SDS-PAGE and HPLC
Bioactivity
3589
Human IL-11
10 μg
RUO
adipogenesis inhibitory factor; AGIF; il 11; IL10RA; Il11; IL-11; IL11 inhibitor; IL-11ra; ILN; Interleukin; interleukin 11; Interleukin11; interleukin-11; MGC2146; Oprelvekin
IL11
Protein
Human
Human IL-11 recombinant protein contains 179 amino acid residues
Carrier-Free
<0.1 ng/μg
E. coli
Recombinant, Ligand
Interleukins
Gibco
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions