missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-1 R3/IL-1 R Acp ELISA Kit
ELISA
$890.00 - $4,420.00
Specifications
| Accession Number | 6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1 R 3; IL 1R3; IL 1 RAcP; IL 1 RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1 R-3; IL1RAcP; IL-1 RAcP; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain |
|---|---|
| Assay Range | 18 to 4,000 pg/mL |
| Assay Sensitivity | 18 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH255RBX5
|
Invitrogen™
EH255RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH255RB
|
Invitrogen™
EH255RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human IL-1 R3/IL-1 R Acp quantitates human IL-1 R3/IL-1 R Acp in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-1 R3/IL-1 R Acp.
This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.Specifications
| 6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1 R 3; IL 1R3; IL 1 RAcP; IL 1 RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1 R-3; IL1RAcP; IL-1 RAcP; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain | |
| 18 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| IL1RAP | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| IL-1 RAcP | |
| 4 hrs 45 mins |
| 18 to 4,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 3556 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |