missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgM Uncoated ELISA Kit with Plates

ELISA

$441.00 - $1,616.00

Specifications

Assay Range 15.6 to 1,000 ng/mL
Assay Sensitivity 15.6 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
501125157
View Documents Promotion Details
Invitrogen™
885062086
10 x 96 Tests
Each for $1,154.00
Only null left
 
501125155
View Documents Promotion Details
Invitrogen™
885062076
20 x 96 Tests
Each for $1,616.00
Only null left
 
501129728
View Documents Promotion Details
Invitrogen™
885062022
2 x 96 Tests
Each for $441.00
Only null left
 
Description

Description

The Human Immunoglobulin M (IgM) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IgM. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications

Specifications

15.6 to 1,000 ng/mL
HRP
Plasma, Serum, Supernatant
ELISA
3507
HRP
Plasma 10 μL, Serum 10 μL, Supernatant 10 μL
Immunoglobulin M
1 hrs 45 mins
15.6 ng/mL
ELISA
Human
Colorimetric Microplate Reader
Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
RUO
2°C to 8°C
Human IgM
24 hrs 30 mins
Videos
SDS
Documents

Documents