missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgG4 ELISA Kit

Catalog No. 501125312
Change view
Click to view available options
Quantity:
2 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125312 96 Tests
501125315 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501125312 Supplier Invitrogen™ Supplier No. BMS2095
Only null left

Includes

Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human total IgG

ELISA

The Human ImmunoGlobulin 4 (Hu IgG4) ELISA quantitates Hu IgG4 in human serum or plasma. The assay will exclusively recognize both natural and recombinant Hu IgG4. Principle of the method The Human IgG4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Human IgG4, also known as human immunoglobulin G4, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the least abundant subclass of IgG in the blood and plays a minor role in defense against pathogens. IgG4 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG4 is also involved in other immune responses, such as immune tolerance and suppression of inflammation. Human IgG4 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.

Specifications

Assay Range 9.4 to 600 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3503
Interassay CV 10.2%
Intraassay CV 4.6
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 20 μL, Serum 20 μL
Storage Requirements 2°C to 8°C
Synonym Immunoglobulin G4
Target Human IgG4
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 30 mins
Show More Show Less