Learn More
Invitrogen™ Human IgG2 Uncoated ELISA Kit with Plates
ELISA
Supplier: Invitrogen™ 885057022
Description
The Human Immunoglobulin G2 (IgG2) Uncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IgG2. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Human IgG2, also known as human immunoglobulin G2, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the second most abundant subclass of IgG in the blood and plays a key role in defense against pathogens. IgG2 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG2 is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG2 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.Chemical Identifiers
| 0.63 to 40 ng/mL | |
| 0.63 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader |
Specifications
| 0.63 to 40 ng/mL | |
| HRP | |
| Plasma, Serum | |
| ELISA | |
| Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86) | |
| 2 x 96 Tests | |
| Plasma 10 μL, Serum 10 μL | |
| Immunoglobulin G2 | |
| 1 hrs 45 mins |
| 0.63 ng/mL | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| Human IgG2 | |
| 24 hrs 30 mins |