missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IGF-I Recombinant Protein

Numéro de catalogue. PHG0071
Encompass
Modifier l'affichage
Click to view available options
Quantité:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PHG0071 100 μg
PHG0078 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. PHG0071 Fournisseur Gibco™ Code fournisseur PHG0071

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Recombinant Protein

Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in water, to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 12 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.

IGF1 (Insulin-like growth factor-1) is structurally and functionally related to insulin but has a much higher growth-promoting activity. A variety of cellular responses are induced by IGF1, including cell proliferation, differentiation, migration, and survival. Further, IGF1 is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types in muscle, bone, and cartilage tissue. IGF1 stimulates glucose transport in rat bone-derived osteoblastic (PyMS) cells and is effective at much lower concentrations than insulin, not only regarding glycogen and DNA synthesis but also with regards to enhancing glucose uptake. In circulation, IGFs are predominantly bound to binding proteins (IGFBPs) which prolong the half-life of the IGFs and play a role in delivering them to target tissues. IGF-I is known as one of the most potent activators of the AKT signaling pathway which is known to be a stimulator of proliferation and an inhibitor of programmed cell death. Moreover, mature human IGF-I is 100% homologous with bovine and porcine proteins. Low levels of IGF1 have been linked to Alzheimer's disease. IGF1 is processed from a precursor, bound by a specific receptor, and secreted. Defects in the IGF1 gene are a cause of insulin-like growth factor 1 deficiency and several transcript variants encoding different isoforms have been found.

Spécifications

Numéro d'enregistrement P05019
À utiliser avec (application) Bioactivity
Formule Protein with no preservative
ID du gène (Entrez) 3479
Poids moléculaire (g/mol) 7.5 kDa
Nom Human IGF-I
Méthode de purification Purified
Quantité 100 μg
Conditions de stockage -20°C
Statut réglementaire RUO
Concentration en endotoxines <0.1 ng/μg
Alias du gène C730016P09Rik; class 1 insulin-like growth factor I preproprotein; class 2 insulin-like growth factor I preproprotein; H-IGF-1; IBP1; IGF; IGF1; Igf-1; IGF1A; IGFI; Igf-I; IGFIa; IGF-IA; IGF-IB; insulin growth factor-1; Insulin like growth factor; insulin like growth factor 1; insulin-like growth factor 1; insulin-like growth factor 1 (somatomedin C); insulin-like growth factor I; insulin-like growth factor I (somatomedin C); insulin-like growth factor IB; mechano growth factor; MGF; M-IGF-1; OTTHUMP00000195084; prepro-IGF-I; prepro-insulin-like growth factor I; R-IGF-1; somatomedin; Somatomedin C; Somatomedin-C
Nom usuel IGF1
Symbole de gène IGF1
Activité biologique ED50 ≤ 2 ng/mL; determined by the dose-dependent proliferation of FDC-P1 cells.
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Séquence GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA
Contenu et stockage -20°C
Classification Carrier-Free
Activité ED50 ≤ 2 ng/mL; determined by the dose-dependent proliferation of FDC-P1 cells.
Dosage biologique proliferation of mouse FDC-P1 cells
Taux d’endotoxine <1 EU/ml
Shipping condition (Condition d'expédition) Wet Ice
Système d’expression E. coli
Famille de protéines Growth Factors & Receptors
Forme de protéine Recombinant, Ligand
Forme Lyophilized
Sous-type de protéine IGFs (Insulin-Like Growth Factors)
Catégorie de recherche Neurobiology, Drug Development, Oncology, Signal Transduction
Grade de qualité ou de pureté ≥98% by SDS-PAGE and HPLC
Afficher plus de résultats Afficher moins de résultats