missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgE ELISA Kit

ELISA

Supplier:  Invitrogen™ BMS2097

 View more versions of this product

Catalog No. 501125320


Only null left

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgE

Description

Description

The Human Immunoglobulin E (Hu IgE) ELISA quantitates Hu IgE in human serum or plasma. The assay will exclusively recognize both natural and recombinant Hu IgE. Principle of the method The Human IgE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.
Specifications

Chemical Identifiers

7.8 to 500 ng/mL
HRP
Plasma, Serum
ELISA
3497
6.6
HRP
Plasma 10 μL, Serum 10 μL
Immunoglobulin E
1 hrs 20 mins
0.5 ng/mL
ELISA
Human
Colorimetric Microplate Reader
5.2%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers
RUO
2°C to 8°C
Human IgE
1 hrs 30 mins

Specifications

7.8 to 500 ng/mL
HRP
Plasma, Serum
ELISA
3497
6.6
HRP
RUO
2°C to 8°C
Human IgE
1 hrs 30 mins
0.5 ng/mL
ELISA
Human
Colorimetric Microplate Reader
5.2%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers
96 Tests
Plasma 10 μL, Serum 10 μL
Immunoglobulin E
1 hrs 20 mins
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions