missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IFN omega ELISA Kit

ELISA

$858.00 - $5,960.00

Specifications

Accession Number P05000
Assay Range 4.7 to 300 pg/mL
Assay Sensitivity 1.5 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IFN-omega
Biotin-Conjugate anti-human IFN-omega monoclonal antibody
Streptavidin-HRP
Human IFN-omega Standard, 6ng/mL
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5018109
View Documents
Invitrogen™
BMS233
96 Tests N/A
Only null left
 
5018112
View Documents
Invitrogen™
BMS233TEN
10 x 96 Tests N/A
Only null left
 
Description

Description

The Human Interferon omega (Hu IFNβ) ELISA quantitates Hu IFNβ in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IFNβ. Principle of the method The Human IFNβ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IFN-omega is a type I interferon that can be induced by virus-infected leukocytes. Members of the type I interferon family, which includes IFN-alpha, IFN-beta, and IFN-omega, signal through the IFNAR-1/IFNAR-2 receptor complex, and exert antiviral and antiproliferative activities. IFN-omega exhibits about 75% sequence homology with IFN-alpha, and contains two conserved disulfide bonds that are necessary for full biological activity.
Specifications

Specifications

P05000
1.5 pg/mL
ELISA
Human
Colorimetric Microplate Reader
IFNW1
4.9
HRP
Plasma 25 μL, Serum 25 μL, Supernatant 25 μL
IFNWP2, Interferon alpha-II-1, Interferon omega-1
1 hrs 20 mins
4.7 to 300 pg/mL
HRP
Plasma, Serum, Supernatant
ELISA
3467
6.8%
Pre-coated 96 well plate, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, SAV-HRP, SAV-HRP, Wash Buffer, Chromogen, Chromogen, Stop Solution
RUO
2°C to 8°C
IFN omega
3 hrs 10 mins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'