missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Human Identification Installation Kit For 3500 Series Genetic Analyzer
Description
- Save time
- Reduce costs
- Increase productivity
Specifications
Specifications
| Description | Human Identification Installation Kit, For 3500 Series Genetic Analyzer |
| For Use With (Equipment) | 3500 Series Genetic Analyzer |
| Product Type | Human Identification Installation Kit |
| Quantity | 1 Kit |
| Size | 1 Kit |
| Source | Human |