missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HYPK (aa 54-120) Control Fragment Recombinant Protein

Catalog No. RP101704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101704 Supplier Invitrogen™ Supplier No. RP101704

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63098. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Huntingtin-interacting protein K (HYPK) has a chaperone-like activity preventing polyglutamine (polyQ) aggregation of HTT. It protects against HTT polyQ-mediated apoptosis in Neuro2a neuronal cells. HYPK is required for optimal NAA10-NAA15 complex-mediated N-terminal acetylation.

Specifications

Accession Number Q9NX55
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 25764
Name Human HYPK (aa 54-120) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias C15orf63; FLJ20431; HSPC136; huntingtin interacting protein K; huntingtin yeast partner K; Huntingtin-interacting protein K; HYPK
Common Name HYPK
Gene Symbol HYPK
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TAMSVIGDRRSREQKAKQEREKELAKVTIKKEDLELIMTEMEISRAAAERSLREHMGNVVEALIALT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less