missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HOOK3 (aa 192-263) Control Fragment Recombinant Protein

Catalog No. RP93890
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93890 Supplier Invitrogen™ Supplier No. RP93890

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55172 (PA5-55172. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.

Specifications

Accession Number Q86VS8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84376
Name Human HOOK3 (aa 192-263) Control Fragment
Quantity 100 μL
Gene Alias 5830454D03Rik; AI317159; E330005F07Rik; hHK3; h-hook3; HK3; hook homolog 3; hook homolog 3 (Drosophila); hook microtubule-tethering protein 3; Hook3; mHK3; protein Hook homolog 3
Common Name HOOK3
Gene Symbol HOOK3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KEEIAQRCHELDMQVAALQEEKSSLLAENQVLMERLNQSDSIEDPNSPAGRRHLQLQTQLEQLQEETFRLEA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less